Assay Details
| Gene Symbol: Prss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008939.2 | NP_032965.1 | 1 - 2 | 721 | 77 | |
| GenBank mRNA | AK132633.1 | - | 1 - 2 | 510 | 77 | |
| BC031429.1 | - | 1 - 2 | 623 | 3665834 | 77 | |
| D89871.1 | - | 1 - 2 | 719 | 77 | ||
| Y13192.1 | - | 1 - 2 | 498 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss12
Genomic Map