Assay Details
| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159561.1 | NP_001153033.1 | 4 - 5 | 601 | 87 | |
| NM_011028.2 | NP_035158.2 | 4 - 5 | 601 | 87 | ||
| GenBank mRNA | AB010883.1 | - | 4 - 5 | 486 | 87 | |
| AK054195.1 | - | 554 | 87 | |||
| AK081308.1 | - | 4 - 5 | 477 | 87 | ||
| AK138541.1 | - | 4 - 5 | 453 | 87 | ||
| BC117713.2 | - | 4 - 5 | 463 | 40088045 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

P2rx6
Genomic Map