Assay Details
| Gene Symbol: Ppp2r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138748.5 | NP_620087.3 | 2 - 3 | 378 | 69 | |
| GenBank mRNA | AK046288.1 | - | 2 - 3 | 182 | 69 | |
| AK049184.1 | - | 2 - 3 | 369 | 69 | ||
| AK150029.1 | - | 2 - 3 | 373 | 69 | ||
| AK160980.1 | - | 2 - 3 | 233 | 69 | ||
| AK179985.1 | - | 1 - 2 | 101 | 69 | ||
| AK185428.1 | - | 1 - 2 | 101 | 69 | ||
| AK207653.1 | - | 1 - 2 | 101 | 69 | ||
| AK212725.1 | - | 1 - 2 | 101 | 69 | ||
| AK219416.1 | - | 1 - 2 | 101 | 69 | ||
| AY035997.1 | - | 2 - 3 | 231 | 69 | ||
| BC006626.1 | - | 2 - 3 | 294 | 3158086 | 69 | |
| BC052852.1 | - | 2 - 3 | 304 | 30058414 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r4
Genomic Map