Assay Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146199.1 | NP_001139671.1 | 2 - 3 | 395 | 93 | |
| NM_011877.2 | NP_036007.2 | 2 - 3 | 530 | 93 | ||
| GenBank mRNA | AK013777.1 | - | 2 - 3 | 532 | 93 | |
| AK030456.1 | - | 2 - 3 | 531 | 93 | ||
| BC014691.1 | - | 2 - 3 | 660 | 93 | ||
| BC049893.1 | - | 1 - 2 | 362 | 93 | ||
| BC098500.1 | - | 2 - 3 | 395 | 30081068 | 93 | |
| D37801.1 | - | 2 - 3 | 479 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn21
Genomic Map