Assay Details
| Gene Symbol: Mrpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025553.4 | NP_079829.1 | 2 - 3 | 311 | 100 | |
| GenBank mRNA | AB049639.1 | - | 2 - 3 | 289 | 100 | |
| AK002797.1 | - | 2 - 3 | 266 | 100 | ||
| AK010329.1 | - | 2 - 3 | 303 | 100 | ||
| AK167247.1 | - | 2 - 3 | 279 | 100 | ||
| BC046750.1 | - | 2 - 3 | 264 | 5692108 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl11
Genomic Map