Assay Details
| Gene Symbol: Mrpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026154.1 | NP_080430.1 | 1 - 2 | 70 | 77 | |
| GenBank mRNA | AK002865.1 | - | 1 - 2 | 70 | 77 | |
| AK007643.1 | - | 1 - 2 | 62 | 77 | ||
| AK078463.1 | - | 1 - 2 | 62 | 77 | ||
| AK150774.1 | - | 1 - 2 | 62 | 77 | ||
| AK171028.1 | - | 1 - 2 | 64 | 77 | ||
| BC016219.1 | - | 1 - 2 | 56 | 4485332 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048330 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180201 | BC016219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180201 | AK150774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180201 | AK171028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180201 | AK002865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180201 | AK007643.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416909 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557068 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143242 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130004 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158944 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180201 | BC016219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180201 | AK150774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180201 | AK171028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180201 | AK002865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180201 | AK007643.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048338 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158944 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143242 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130004 | - | - |
| Whole Mouse Genome G4122A | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | BC016219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK150774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK002865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK007643.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384640 | AK078463.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130886 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143242 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158944 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09737354 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09737356 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09665406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09737356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09280564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09699341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09699329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048345 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380737 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418112_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380732 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600899 | - | - |
| Mouse Genome 430 2.0 Array | 1418112_at | - | - |
| Mouse Genome 430 2.0 Array | 1459966_at | - | - |
| Mouse Genome 430 2.0 Array | 1418112_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459966_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107732_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107732_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014074.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014074.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014074.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014070.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014070.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014070.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014078.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014078.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014078.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014077.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014077.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014077.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014079.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014079.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014079.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014072.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014072.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014072.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014075.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014075.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014075.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014076.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014076.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014076.mm.1 | TC1100001529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014080.mm.1 | NM_026154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014080.mm.1 | NM_026154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014080.mm.1 | TC1100001529.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418112_at | - | - |
| Mouse Expression Set 430 | 1418112_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418112_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418112_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418112_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418112_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 331417 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610057 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610057 | - | - |
| mouse-6 v1.1 (Illumina) | scl40969.4.1_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40969.4.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2822622 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2822621 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2822622 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2822621 | - | - |

Mrpl10
Genomic Map