Assay Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077705.2 | NP_001071173.1 | 10 - 11 | 1410 | 104 | |
| NM_013545.3 | NP_038573.2 | 10 - 11 | 1408 | 104 | ||
| GenBank mRNA | AK132509.1 | - | 10 - 11 | 1327 | 104 | |
| AK150509.1 | - | 10 - 11 | 1315 | 104 | ||
| AK151077.1 | - | 10 - 11 | 1329 | 104 | ||
| AK151623.1 | - | 10 - 11 | 1332 | 104 | ||
| AK159686.1 | - | 10 - 11 | 1367 | 104 | ||
| AK159833.1 | - | 10 - 11 | 1360 | 104 | ||
| AK167923.1 | - | 10 - 11 | 1362 | 104 | ||
| AK169804.1 | - | 10 - 11 | 1410 | 104 | ||
| BC012660.1 | - | 10 - 11 | 1353 | 4010462 | 104 | |
| M68902.1 | - | 10 - 11 | 1360 | 104 | ||
| M90389.1 | - | 10 - 11 | 1310 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn6
Genomic Map