Assay Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077705.2 | NP_001071173.1 | 14 - 15 | 1873 | 70 | |
| NM_013545.3 | NP_038573.2 | 14 - 15 | 1871 | 70 | ||
| GenBank mRNA | AK132509.1 | - | 14 - 15 | 1790 | 70 | |
| AK150509.1 | - | 14 - 15 | 1778 | 70 | ||
| AK151077.1 | - | 14 - 15 | 1792 | 70 | ||
| AK151623.1 | - | 14 - 15 | 1795 | 70 | ||
| AK167923.1 | - | 14 - 15 | 1825 | 70 | ||
| AK169804.1 | - | 14 - 15 | 1873 | 70 | ||
| BC012660.1 | - | 14 - 15 | 1816 | 4010462 | 70 | |
| M68902.1 | - | 14 - 15 | 1823 | 70 | ||
| M90389.1 | - | 14 - 15 | 1773 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn6
Genomic Map