Assay Details
| Gene Symbol: Pet2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008821.2 | NP_032847.2 | 11 - 12 | 1342 | 115 | |
| GenBank mRNA | BC138080.1 | - | 10 - 11 | 1163 | 8861099 | 115 |
| M95564.1 | - | 11 - 12 | 1347 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138080.1, M95564.1, NM_008821.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960112 | NM_008821.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960112 | M95564.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547036 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111257 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171618 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960112 | NM_008821.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960112 | M95564.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32521217 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171618 | - | - |
| Whole Mouse Genome G4122A | A_52_P423238 | NM_008821.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423238 | BC138080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423238 | M95564.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125869 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108137 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32521247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14944604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15060725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15107254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14886618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14581094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14862321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15051365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14724794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14581099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15051354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15051360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15060723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32521247 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10605639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605642 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421566_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605633 | - | - |
| Mouse Genome 430 2.0 Array | 1421566_at | - | - |
| Mouse Genome 430 2.0 Array | 1421566_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18630_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18630_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa10570_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00016316.mm.1 | NM_008821.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016316.mm.1 | NM_008821.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016316.mm.1 | TC0X00002600.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421566_at | - | - |
| Mouse Expression Set 430 | 1421566_PM_at | - | - |
| Murine Genome U74v2 Set | 96597_at | - | - |
| Murine Genome U74v2 Set | 96597_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421566_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421566_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421566_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421566_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 430395 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5390100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5390100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005390100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2619442 | - | - |
| mouse-6 v1.1 (Illumina) | scl018630.1_158-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5390100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl018630.1_158-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5390100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2821371 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2821371 | - | - |

Pet2
Genomic Map