Assay Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011206.2 | NP_035336.2 | 9 - 10 | 836 | 69 | |
| GenBank mRNA | AK144353.1 | - | 9 - 10 | 818 | 69 | |
| U35124.1 | - | 9 - 10 | 810 | 69 | ||
| U49853.1 | - | 9 - 10 | 776 | 69 | ||
| U52523.1 | - | 9 - 10 | 824 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn18
Genomic Map