Assay Details
| Gene Symbol: Gkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025466.1 | NP_079742.1 | 1 - 2 | 48 | 73 | |
| GenBank mRNA | AK008622.1 | - | 1 - 2 | 48 | 73 | |
| AK008640.1 | - | 1 - 2 | 45 | 73 | ||
| AK008641.1 | - | 1 - 2 | 48 | 73 | ||
| AK008647.1 | - | 1 - 2 | 45 | 73 | ||
| AK008722.1 | - | 1 - 2 | 46 | 73 | ||
| AK008745.1 | - | 1 - 2 | 48 | 73 | ||
| AK008933.1 | - | 1 - 2 | 50 | 73 | ||
| AK008956.1 | - | 1 - 2 | 45 | 73 | ||
| AK008990.1 | - | 1 - 2 | 50 | 73 | ||
| AK009145.1 | - | 1 - 2 | 46 | 73 | ||
| AK019050.1 | - | 1 - 2 | 49 | 73 | ||
| AK028136.1 | - | 1 - 2 | 44 | 73 | ||
| AY456960.1 | - | 1 - 2 | 48 | 73 | ||
| BC132540.1 | - | 1 - 2 | 38 | 40130817 | 73 | |
| BC132542.1 | - | 1 - 2 | 38 | 40130819 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008622.1,AK008641.1,AK008647.1,AK008722.1,AK008745.1,AK008933.1,AK008990.1,AK009145.1,AK019050.1,AK028136.1,AY456960.1,BC132540.1,BC132542.1,NM_025466.1 transcripts.
Set Membership: |
|

Gkn1
Genomic Map