Assay Details
| Gene Symbol: Dlg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177778.2 | NP_001171249.1 | 16 - 17 | 2370 | 79 | |
| NM_001177779.1 | NP_001171250.1 | 10 - 11 | 1367 | 79 | ||
| NM_001177780.1 | NP_001171251.1 | 8 - 9 | 759 | 79 | ||
| NM_001290402.1 | NP_001277331.1 | 15 - 16 | 2316 | 79 | ||
| NM_016747.4 | NP_058027.1 | 17 - 18 | 2412 | 79 | ||
| GenBank mRNA | AK122474.1 | - | 16 - 17 | 2369 | 79 | |
| AK145815.1 | - | 10 - 11 | 1122 | 79 | ||
| AK148464.1 | - | 8 - 9 | 739 | 79 | ||
| AK159655.1 | - | 8 - 9 | 747 | 79 | ||
| AK167365.1 | - | 8 - 9 | 746 | 79 | ||
| BC060713.1 | - | 15 - 16 | 2444 | 79 | ||
| BC094368.1 | - | 15 - 16 | 2311 | 6842105 | 79 | |
| D87117.1 | - | 17 - 18 | 2386 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlg3
Genomic Map