Assay Details
| Gene Symbol: Ppp2r3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163415.1 | NP_001156887.1 | 900 | 64 | ||
| NM_011154.2 | NP_035284.2 | 666 | 64 | |||
| NR_028110.2 | 666 | 64 | ||||
| NR_102716.1 | 666 | 64 | ||||
| GenBank mRNA | AF050165.1 | - | 663 | 64 | ||
| AK054122.1 | - | 666 | 64 | |||
| AK143212.1 | - | 454 | 64 | |||
| BC027415.1 | - | 643 | 4235453 | 64 | ||
| BC059852.1 | - | 931 | 6402777 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143212.1 transcripts.
Set Membership: |
|

Ppp2r3d
Genomic Map