Assay Details
| Gene Symbol: Polr3gl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027241.4 | NP_081517.2 | 2 - 3 | 265 | 79 | |
| GenBank mRNA | AK008069.1 | - | 2 - 3 | 231 | 79 | |
| BC027083.1 | - | 2 - 3 | 215 | 5357914 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr3gl
Genomic Map