Assay Details
| Gene Symbol: Polr3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029763.3 | NP_084039.2 | 5 - 6 | 544 | 123 | |
| GenBank mRNA | AK013897.1 | - | 5 - 6 | 531 | 123 | |
| AK029286.1 | - | 5 - 6 | 545 | 123 | ||
| AK036166.1 | - | 5 - 6 | 541 | 123 | ||
| AK076538.1 | - | 5 - 6 | 571 | 123 | ||
| AK144679.1 | - | 5 - 6 | 542 | 123 | ||
| BC009159.1 | - | 5 - 6 | 533 | 3602180 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr3f
Genomic Map