Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025903.2 | NP_080179.1 | 2 - 3 | 347 | 77 | |
| GenBank mRNA | AK007687.1 | - | 2 - 3 | 348 | 77 | |
| AK149496.1 | - | 2 - 3 | 328 | 77 | ||
| AK156625.1 | - | 2 - 3 | 341 | 77 | ||
| AK165967.1 | - | 2 - 3 | 340 | 77 | ||
| AK166130.1 | - | 2 - 3 | 415 | 77 | ||
| AK166258.1 | - | 2 - 3 | 305 | 77 | ||
| AY298860.1 | - | 2 - 3 | 176 | 77 | ||
| BC023486.1 | - | 2 - 3 | 295 | 77 | ||
| BC037434.1 | - | 2 - 3 | 303 | 2651172 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd2
Genomic Map