Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025903.2 | NP_080179.1 | 8 - 9 | 1048 | 68 | |
| GenBank mRNA | AK002702.1 | - | 2 - 3 | 273 | 68 | |
| AK007687.1 | - | 8 - 9 | 1049 | 68 | ||
| AK149496.1 | - | 8 - 9 | 1029 | 68 | ||
| AK156625.1 | - | 7 - 8 | 1124 | 68 | ||
| AK165967.1 | - | 8 - 9 | 1042 | 68 | ||
| AK166130.1 | - | 8 - 9 | 1116 | 68 | ||
| AK166258.1 | - | 8 - 9 | 1006 | 68 | ||
| AY298860.1 | - | 8 - 9 | 877 | 68 | ||
| BC023486.1 | - | 8 - 9 | 995 | 68 | ||
| BC037434.1 | - | 8 - 9 | 1004 | 2651172 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd2
Genomic Map