Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025903.2 | NP_080179.1 | 11 - 12 | 1421 | 67 | |
| GenBank mRNA | AK002702.1 | - | 5 - 6 | 646 | 67 | |
| AK007687.1 | - | 11 - 12 | 1422 | 67 | ||
| AK149496.1 | - | 11 - 12 | 1402 | 67 | ||
| AK156625.1 | - | 10 - 11 | 1497 | 67 | ||
| AK165967.1 | - | 11 - 12 | 1415 | 67 | ||
| AK166130.1 | - | 11 - 12 | 1489 | 67 | ||
| AK166258.1 | - | 11 - 12 | 1379 | 67 | ||
| AY298860.1 | - | 11 - 12 | 1250 | 67 | ||
| BC023486.1 | - | 11 - 12 | 1368 | 67 | ||
| BC037434.1 | - | 11 - 12 | 1377 | 2651172 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809753 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809757 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809752 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809756 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809755 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809754 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809758 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809760 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | BC037434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | AK156625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | NM_025903.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | AK007687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | BC037434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | AK156625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | NM_025903.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172251 | AK007687.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274333 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106252 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | BC037434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | AK156625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | NM_025903.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | AK007687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | BC037434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | AK156625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | NM_025903.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172251 | AK007687.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809764 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106252 | - | - |
| Whole Mouse Genome G4122A | A_52_P322006 | BC023486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322006 | BC037434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322006 | AK166130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322006 | AK166258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322006 | AY298860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | BC037434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | AK156625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | NM_025903.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | AK007687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | BC037434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | AK156625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | NM_025903.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172251 | AK007687.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116456 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106252 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08191113 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809760 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809754 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809755 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809758 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809756 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809757 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809752 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26809753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08191125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07792771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08191106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588711 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588710 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451016_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588707 | - | - |
| Mouse Genome 430 2.0 Array | 1447393_at | - | - |
| Mouse Genome 430 2.0 Array | 1451016_at | - | - |
| Mouse Genome 430 2.0 Array | 1451016_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447393_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15983_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15983_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900010862.mm.1 | NM_025903.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010862.mm.1 | NM_025903.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010862.mm.1 | TC0900001377.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451016_at | - | - |
| Mouse Expression Set 430 | 1451016_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451016_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451016_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451016_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451016_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 770176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736761 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736761 | - | - |
| mouse-6 v1.1 (Illumina) | scl36492.10.1_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770176 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36492.10.1_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770176 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2736762 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2736762 | - | - |

Ifrd2
Genomic Map