Assay Details
| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038627.1 | NP_001033716.1 | 15 - 16 | 2084 | 77 | |
| NM_029438.3 | NP_083714.3 | 15 - 16 | 2084 | 77 | ||
| GenBank mRNA | AK015264.1 | - | 11 - 12 | 1483 | 77 | |
| AK034736.1 | - | 6 - 7 | 682 | 77 | ||
| AK122534.1 | - | 4 - 5 | 459 | 77 | ||
| AK154491.1 | - | 15 - 16 | 2085 | 77 | ||
| BC029097.1 | - | 15 - 16 | 2112 | 3660965 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smurf1
Genomic Map