Assay Details
| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 6 - 7 | 1232 | 86 | |
| GenBank mRNA | AK030039.1 | - | 6 - 7 | 1236 | 86 | |
| AK042071.1 | - | 4 - 5 | 789 | 86 | ||
| AK047545.1 | - | 6 - 7 | 1225 | 86 | ||
| AK053356.1 | - | 6 - 7 | 1223 | 86 | ||
| AK085666.1 | - | 6 - 7 | 1225 | 86 | ||
| AK128956.1 | - | 6 - 7 | 1224 | 86 | ||
| AK154181.1 | - | 6 - 7 | 1223 | 86 | ||
| AK154729.1 | - | 6 - 7 | 1214 | 86 | ||
| BC110697.1 | - | 6 - 7 | 1208 | 30356222 | 86 | |
| BC117939.1 | - | 6 - 7 | 1225 | 86 | ||
| BC117940.1 | - | 6 - 7 | 1225 | 40097816 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polrmt
Genomic Map