Assay Details
| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 9 - 10 | 1796 | 58 | |
| GenBank mRNA | AK042071.1 | - | 7 - 8 | 1353 | 58 | |
| AK047545.1 | - | 9 - 10 | 1789 | 58 | ||
| AK053356.1 | - | 9 - 10 | 1787 | 58 | ||
| AK085666.1 | - | 9 - 10 | 1789 | 58 | ||
| AK128956.1 | - | 9 - 10 | 1788 | 58 | ||
| AK154181.1 | - | 9 - 10 | 1787 | 58 | ||
| AK154729.1 | - | 9 - 10 | 1778 | 58 | ||
| BC110697.1 | - | 9 - 10 | 1772 | 30356222 | 58 | |
| BC117939.1 | - | 9 - 10 | 1789 | 58 | ||
| BC117940.1 | - | 9 - 10 | 1789 | 40097816 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polrmt
Genomic Map