Assay Details
| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013753.2 | NP_038781.2 | 15 - 16 | 2175 | 67 | |
| GenBank mRNA | AK088577.1 | - | 15 - 16 | 2011 | 67 | |
| AK146489.1 | - | 15 - 16 | 2120 | 67 | ||
| BC137695.1 | - | 14 - 15 | 1939 | 8860714 | 67 | |
| BC144846.1 | - | 14 - 15 | 1948 | 9053394 | 67 | |
| BC144847.1 | - | 14 - 15 | 1945 | 9053395 | 67 | |
| X99384.1 | - | 14 - 15 | 1960 | 67 | ||
More Information
Set Membership: |
|

Pald1
Genomic Map