| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00016332.mm.1 |
NM_008821.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016332.mm.1 |
NM_008821.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016332.mm.1 |
TC0X00002600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00016330.mm.1 |
NM_008821.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016330.mm.1 |
NM_008821.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016330.mm.1 |
TC0X00002600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00016327.mm.1 |
NM_008821.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016327.mm.1 |
NM_008821.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016327.mm.1 |
TC0X00002600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00016331.mm.1 |
NM_008821.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016331.mm.1 |
NM_008821.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016331.mm.1 |
TC0X00002600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00016329.mm.1 |
NM_008821.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016329.mm.1 |
NM_008821.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00016329.mm.1 |
TC0X00002600.mm.1 |
MTA TC |