Assay Details
| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011945.2 | NP_036075.2 | 15 - 16 | 3780 | 95 | |
| GenBank mRNA | AF117340.1 | - | 15 - 16 | 3780 | 95 | |
| L13103.1 | - | 5 - 6 | 1789 | 95 | ||
| U23470.1 | - | 2 - 3 | 1381 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k1
Genomic Map