Assay Details
| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011945.2 | NP_036075.2 | 18 - 19 | 4217 | 92 | |
| GenBank mRNA | AF117340.1 | - | 18 - 19 | 4217 | 92 | |
| L13103.1 | - | 8 - 9 | 2226 | 92 | ||
| U23470.1 | - | 5 - 6 | 1818 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k1
Genomic Map