Assay Details
| Gene Symbol: Ppef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011147.1 | NP_035277.1 | 11 - 12 | 1372 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppef1| Gene Symbol: Ppef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011147.1 | NP_035277.1 | 11 - 12 | 1372 | 84 | |
Set Membership: |
|