| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00020482.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020482.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020482.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020488.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020488.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020488.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020491.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020491.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020491.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020483.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020483.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020483.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020485.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020485.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020485.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020492.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020492.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020492.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020484.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020484.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020484.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020489.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020489.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020489.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020486.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020486.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020486.mm.1 |
TC0X00003276.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00020487.mm.1 |
NM_011147.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020487.mm.1 |
NM_011147.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00020487.mm.1 |
TC0X00003276.mm.1 |
MTA TC |