Assay Details
| Gene Symbol: Ppp1r9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181595.3 | NP_853626.1 | 6 - 7 | 2242 | 74 | |
| GenBank mRNA | AB091828.1 | - | 6 - 7 | 2242 | 74 | |
| AK029737.1 | - | 6 - 7 | 1008 | 74 | ||
| AK138313.1 | - | 6 - 7 | 2171 | 74 | ||
| AY308065.1 | - | 5 - 6 | 1917 | 74 | ||
| BC053748.1 | - | 6 - 7 | 2130 | 6331919 | 74 | |
More Information
Set Membership: |
|

Ppp1r9a
Genomic Map