| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500001935.mm.1 |
NM_001039368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001935.mm.1 |
TC1500000262.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500001936.mm.1 |
NM_001039368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001936.mm.1 |
NM_023127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001936.mm.1 |
TC1500000262.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500001933.mm.1 |
NM_001039368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001933.mm.1 |
NM_023127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001933.mm.1 |
TC1500000262.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500001937.mm.1 |
NM_001039368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001937.mm.1 |
NM_023127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001937.mm.1 |
TC1500000262.mm.1 |
MTA TC |