Assay Details
| Gene Symbol: Polr2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039368.2 | NP_001034457.1 | 3 - 4 | 359 | 84 | |
| NM_023127.3 | NP_075616.2 | 3 - 4 | 227 | 84 | ||
| GenBank mRNA | AK003023.1 | - | 3 - 4 | 209 | 84 | |
| AK008136.1 | - | 3 - 4 | 316 | 84 | ||
| AK008287.1 | - | 3 - 4 | 317 | 84 | ||
| AK018914.1 | - | 3 - 4 | 186 | 84 | ||
| BC028543.1 | - | 175 | 3468916 | 84 | ||
| BY100585.1 | - | 3 - 4 | 222 | 84 | ||
| CR516049.1 | - | 1 - 2 | 84 | |||
| S63758.1 | - | 3 - 4 | 288 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137024 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137021 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137020 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996872 | S63758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996872 | BY100585.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996872 | NM_001039368.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996872 | NM_023127.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996872 | AK003023.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414692 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169659 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996872 | S63758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996872 | BY100585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996872 | NM_001039368.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996872 | NM_023127.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996872 | AK003023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214449 | S63758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214449 | BY100585.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214449 | NM_001039368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214449 | NM_023127.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214449 | AK003023.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30137039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169659 | - | - |
| Whole Mouse Genome G4122A | A_51_P214449 | S63758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214449 | BY100585.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214449 | NM_001039368.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214449 | NM_023127.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214449 | AK003023.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129264 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111942 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169659 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12001313 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11927029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137020 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137021 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137024 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30137023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30137107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12001313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12080304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12092719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30137064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10423744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10599347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423743 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452596_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599346 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10423742 | - | - |
| Mouse Genome 430 2.0 Array | 1452596_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452596_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500001938.mm.1 | NM_001039368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001938.mm.1 | NM_023127.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001938.mm.1 | TC1500000262.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500001939.mm.1 | NM_001039368.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001939.mm.1 | NM_023127.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001939.mm.1 | TC1500000262.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452596_at | - | - |
| Mouse Expression Set 430 | 1452596_PM_at | - | - |
| Murine Genome U74v2 Set | 95003_PM_at | - | - |
| Murine Genome U74v2 Set | 95003_at | - | - |
| Murine Genome U74v2 Set | 170286_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170286_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452596_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452596_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452596_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452596_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 868644 | - | - |
| ABI Mouse Genome Survey Microarray | 904383 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3870400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102940017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3870400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003870400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736071 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736071 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2436809 | - | - |
| mouse-6 v1.1 (Illumina) | scl47983.1.50_24-S | - | - |
| mouse-6 v1.1 (Illumina) | scl017749.3_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3870400 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102940017 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017749.3_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102940017 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2736071 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2736071 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017749.3_11-S | - | - |

Polr2k
Genomic Map