Assay Details
| Gene Symbol: Prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133351.3 | NP_579929.1 | 1 - 2 | 315 | 64 | |
| GenBank mRNA | AB038244.1 | - | 1 - 2 | 296 | 64 | |
| AF188613.1 | - | 1 - 2 | 229 | 64 | ||
| AF378085.2 | - | 1 - 2 | 306 | 64 | ||
| AK010640.1 | - | 1 - 2 | 198 | 64 | ||
| AK078696.1 | - | 1 - 2 | 267 | 64 | ||
| AY335911.1 | - | 2 - 3 | 163 | 64 | ||
| BC003851.1 | - | 1 - 2 | 289 | 3600399 | 64 | |
| CB600036.1 | - | 1 - 2 | 318 | 64 | ||
More Information
Set Membership: |
|

Prss8
Genomic Map