Assay Details
| Gene Symbol: Gm15520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077358.1 | - | 5 - 6 | 862 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurotrimin | Ntm |
6230410L23Rik, B230210G24Rik, Hnt, Igdcc2, R75390 |
Anti-sense | Chr. 9 - 28994923 - 29963486 on NCBI Build GRCm38 | |
| Downstream | sorting nexin 19 | Snx19 |
3526401K03Rik, AI195321, AI848208, mKIAA0254 |
1.4mb | Sense | Chr. 9 - 30427329 - 30466726 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15520
Genomic Map