Assay Details
| Gene Symbol: Pstpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011193.2 | NP_035323.2 | 3 - 4 | 645 | 83 | |
| GenBank mRNA | AK037936.1 | - | 3 - 4 | 647 | 83 | |
| AK088150.1 | - | 3 - 4 | 642 | 83 | ||
| BC096761.1 | - | 3 - 4 | 425 | 30040955 | 83 | |
| U87814.1 | - | 3 - 4 | 485 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pstpip1
Genomic Map