Assay Details
| Gene Symbol: Clpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044389.2 | NP_001037854.1 | 6 - 7 | 1083 | 117 | |
| NM_011802.3 | NP_035932.2 | 7 - 8 | 1125 | 117 | ||
| GenBank mRNA | AF134983.1 | - | 7 - 8 | 1076 | 117 | |
| AJ276991.1 | - | 7 - 8 | 1007 | 117 | ||
| AK087848.1 | - | 7 - 8 | 1079 | 117 | ||
| BC061153.1 | - | 6 - 7 | 1041 | 6593118 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clpx
Genomic Map