Assay Details
| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013373.2 | NP_001013391.2 | 9 - 10 | 1264 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss13| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013373.2 | NP_001013391.2 | 9 - 10 | 1264 | 82 | |
Set Membership: |
|