Assay Details
| Gene Symbol: Mrpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025430.3 | NP_079706.1 | 3 - 4 | 404 | 81 | |
| GenBank mRNA | AK004385.1 | - | 3 - 4 | 397 | 81 | |
| AK010223.1 | - | 3 - 4 | 393 | 81 | ||
| AK010369.1 | - | 3 - 4 | 390 | 81 | ||
| AK012807.1 | - | 3 - 4 | 391 | 81 | ||
| AK039187.1 | - | 3 - 4 | 384 | 81 | ||
| BB615775.1 | - | 3 - 4 | 404 | 81 | ||
| BC028750.1 | - | 3 - 4 | 396 | 1245736 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24538439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24538437 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24538438 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8617 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152390 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141156 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24538432 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05937185 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141156 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152390 | - | - |
| Whole Mouse Genome G4122A | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P351697 | AK039187.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P351697 | NM_025430.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P351697 | AK004385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P223407 | AK039187.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P223407 | AK135623.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P223407 | NM_025430.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590269 | BC028750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590269 | AK012807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590269 | AK039187.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590269 | AK010369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590269 | NM_025430.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112099 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141156 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152390 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05104828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05543820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05222998 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05222999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05104826 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05421613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05104824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05505850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24538438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24538439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24538437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24538392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05222998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05428301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05510450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05589752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05551593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05589751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05510449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05783198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05589749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05336090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05754807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05174084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05605266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05510446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24538421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05222999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545344 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545343 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419362_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419363_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440989_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545339 | - | - |
| Mouse Genome 430 2.0 Array | 1419362_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419363_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1419362_at | - | - |
| Mouse Genome 430 2.0 Array | 1440989_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440989_at | - | - |
| Mouse Genome 430 2.0 Array | 1419363_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66223_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66223_PM_at | - | - |
| Mouse Expression Set 430 | 1419363_PM_a_at | - | - |
| Mouse Expression Set 430 | 1419362_at | - | - |
| Mouse Expression Set 430 | 1419363_a_at | - | - |
| Mouse Expression Set 430 | 1419362_PM_at | - | - |
| Mouse Expression Set 430 | 1440989_at | - | - |
| Mouse Expression Set 430 | 1440989_PM_at | - | - |
| Murine Genome U74v2 Set | 98904_at | - | - |
| Murine Genome U74v2 Set | 98904_PM_at | - | - |
| Murine Genome U74v2 Set | 167104_PM_at | - | - |
| Murine Genome U74v2 Set | 167104_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419363_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419362_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419363_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419362_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419362_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419362_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419363_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419363_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440989_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440989_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 831702 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105890324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105890324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990671 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753924 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2753924 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228027 | - | - |
| mouse-6 v1.1 (Illumina) | scl0066223.2_60-S | - | - |
| mouse-6 v1.1 (Illumina) | scl28878.4_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105890324 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066223.2_60-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990671 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105890324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2753924 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2753924 | - | - |

Mrpl35
Genomic Map