Assay Details
| Gene Symbol: Rimklb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027664.1 | NP_081940.1 | 3 - 4 | 671 | 88 | |
| GenBank mRNA | AK016936.1 | - | 2 - 3 | 397 | 88 | |
| AK039398.1 | - | 2 - 3 | 345 | 88 | ||
| AK173135.1 | - | 4 - 5 | 681 | 88 | ||
| BC052078.1 | - | 3 - 4 | 643 | 6402765 | 88 | |
More Information
Set Membership: |
|

Rimklb
Genomic Map