Assay Details
| Gene Symbol: Pet2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008821.2 | NP_032847.2 | 2 - 3 | 258 | 71 | |
| GenBank mRNA | BC138080.1 | - | 1 - 2 | 62 | 8861099 | 71 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138080.1, NM_008821.2 transcripts.
Set Membership: |
|

Pet2
Genomic Map