Assay Details
| Gene Symbol: Pet2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008821.2 | NP_032847.2 | 8 - 9 | 972 | 69 | |
| GenBank mRNA | BC138080.1 | - | 7 - 8 | 793 | 8861099 | 69 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138080.1, NM_008821.2 transcripts.
Set Membership: |
|

Pet2
Genomic Map