Assay Details
| Gene Symbol: Crcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007761.2 | NP_031787.1 | 2 - 3 | 136 | 87 | |
| GenBank mRNA | AF028242.1 | - | 2 - 3 | 126 | 87 | |
| AF118271.1 | - | 2 - 3 | 136 | 87 | ||
| AK075587.1 | - | 2 - 3 | 131 | 86 | ||
| BC012676.1 | - | 2 - 3 | 70 | 3989860 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF028242.1, AF118271.1, BC012676.1, NM_007761.2 transcripts.
Set Membership: |
|

Crcp
Genomic Map