Assay Details
| Gene Symbol: Crcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007761.2 | NP_031787.1 | 3 - 4 | 237 | 78 | |
| GenBank mRNA | AF028242.1 | - | 3 - 4 | 227 | 78 | |
| AF118271.1 | - | 3 - 4 | 237 | 78 | ||
| AK075587.1 | - | 3 - 4 | 232 | 78 | ||
| BC012676.1 | - | 3 - 4 | 171 | 3989860 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crcp
Genomic Map