Assay Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF201963.1 | - | 162 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF201963.1 transcripts.
Set Membership: |
|

Crlf2
Genomic Map