Assay Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164735.1 | NP_001158207.1 | 5 - 6 | 762 | 153 | |
| GenBank mRNA | AF201963.1 | - | 5 - 6 | 813 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crlf2
Genomic Map