Assay Details
| Gene Symbol: Polr2j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011293.2 | NP_035423.2 | 3 - 4 | 382 | 68 | |
| GenBank mRNA | AK158610.1 | - | 3 - 4 | 383 | 68 | |
| AK158644.1 | - | 3 - 4 | 372 | 68 | ||
| BC042939.1 | - | 3 - 4 | 339 | 6476299 | 68 | |
| DV065741.1 | - | 416 | 68 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119517 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996643 | AK167868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996643 | AK158644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996643 | BC042939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996643 | DV065741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996643 | NM_011293.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3707 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137728 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996643 | AK167868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996643 | AK158644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996643 | BC042939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996643 | DV065741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996643 | NM_011293.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326086 | AK158644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326086 | BC042939.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326086 | D85818.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326086 | DV065741.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326086 | NM_011293.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119521 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137728 | - | - |
| Whole Mouse Genome G4122A | A_51_P326086 | AK158644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P326086 | BC042939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P326086 | D85818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P326086 | DV065741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P326086 | NM_011293.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112304 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04677941 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04937763 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04937765 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04677936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04677940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04677939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04677937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04281023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04190792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04916955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04677940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04592104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04677936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04445793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04281022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04547898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526484 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526483 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440065_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417720_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526482 | - | - |
| Mouse Genome 430 2.0 Array | 1417720_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440065_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417720_at | - | - |
| Mouse Genome 430 2.0 Array | 1440065_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20022_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20022_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500014492.mm.1 | NM_011293.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014492.mm.1 | NM_011293.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014492.mm.1 | TC0500001598.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417720_PM_at | - | - |
| Mouse Expression Set 430 | 1417720_at | - | - |
| Mouse Expression Set 430 | 1440065_at | - | - |
| Mouse Expression Set 430 | 1440065_PM_at | - | - |
| Murine Genome U74v2 Set | 100461_PM_at | - | - |
| Murine Genome U74v2 Set | 100461_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417720_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417720_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417720_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417720_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440065_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440065_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7100372 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007100372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239525 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239525 | - | - |
| mouse-6 v1.1 (Illumina) | scl27119.4.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7100372 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27119.4.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100372 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239525 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239525 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl27119.4.1_2-S | - | - |

Polr2j
Genomic Map