Assay Details
| Gene Symbol: N4bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024917.1 | NP_001020088.1 | 14 - 15 | 4811 | 137 | |
| GenBank mRNA | AK136182.1 | - | 4 - 5 | 1958 | 137 | |
| AK173167.1 | - | 12 - 13 | 4522 | 137 | ||
| BC046458.1 | - | 5 - 6 | 2791 | 137 | ||
| BC054549.1 | - | 14 - 15 | 4811 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

N4bp2
Genomic Map