Assay Details
| Gene Symbol: N4bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024917.1 | NP_001020088.1 | 17 - 18 | 5259 | 66 | |
| GenBank mRNA | AK136182.1 | - | 7 - 8 | 2406 | 66 | |
| CK637074.1 | - | 2 - 3 | 226 | 66 | ||
| CX769144.1 | - | 1 - 2 | 204 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

N4bp2
Genomic Map