Assay Details
| Gene Symbol: Mtus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029920.7 | NP_084196.4 | 10 - 11 | 4392 | 106 | |
| GenBank mRNA | AB093263.1 | - | 9 - 10 | 3266 | 106 | |
| AK021162.1 | - | 4 - 5 | 594 | 106 | ||
| AK147457.1 | - | 10 - 11 | 4393 | 106 | ||
| BC055016.1 | - | 4 - 5 | 635 | 106 | ||
| BC085194.1 | - | 10 - 11 | 3738 | 106 | ||
| BC150942.1 | - | 10 - 11 | 3826 | 9087853 | 106 | |
More Information
Set Membership: |
|

Mtus2
Genomic Map