Assay Details
| Gene Symbol: Ppp2r3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163415.1 | NP_001156887.1 | 1756 | 120 | ||
| NM_011154.2 | NP_035284.2 | 1522 | 120 | |||
| NR_028110.2 | 1864 | 120 | ||||
| NR_102716.1 | 2013 | 120 | ||||
| GenBank mRNA | AF050165.1 | - | 1519 | 120 | ||
| AK054122.1 | - | 1864 | 120 | |||
| AK143212.1 | - | 1799 | 120 | |||
| BC027415.1 | - | 1499 | 4235453 | 120 | ||
| BC059852.1 | - | 1787 | 6402777 | 120 | ||
| BC095938.1 | - | 646 | 120 | |||
| BC096544.1 | - | 1202 | 6834351 | 120 | ||
More Information
Set Membership: |
|

Ppp2r3d
Genomic Map