Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025903.2 | NP_080179.1 | 10 - 11 | 1318 | 60 | |
| GenBank mRNA | AK002702.1 | - | 4 - 5 | 543 | 60 | |
| AK007687.1 | - | 10 - 11 | 1319 | 60 | ||
| AK149496.1 | - | 10 - 11 | 1299 | 60 | ||
| AK156625.1 | - | 9 - 10 | 1394 | 60 | ||
| AK165967.1 | - | 10 - 11 | 1312 | 60 | ||
| AK166130.1 | - | 10 - 11 | 1386 | 60 | ||
| AK166258.1 | - | 10 - 11 | 1276 | 60 | ||
| AY298860.1 | - | 10 - 11 | 1147 | 60 | ||
| BC023486.1 | - | 10 - 11 | 1265 | 60 | ||
| BC037434.1 | - | 10 - 11 | 1274 | 2651172 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd2
Genomic Map