Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025903.2 | NP_080179.1 | 4 - 5 | 558 | 60 | |
| GenBank mRNA | AK007687.1 | - | 4 - 5 | 559 | 60 | |
| AK149496.1 | - | 4 - 5 | 539 | 60 | ||
| AK165967.1 | - | 4 - 5 | 552 | 60 | ||
| AK166130.1 | - | 4 - 5 | 626 | 60 | ||
| AK166258.1 | - | 4 - 5 | 516 | 60 | ||
| AK178654.1 | - | 1 - 2 | 29 | 60 | ||
| AY298860.1 | - | 4 - 5 | 387 | 60 | ||
| BC023486.1 | - | 4 - 5 | 506 | 60 | ||
| BC037434.1 | - | 4 - 5 | 514 | 2651172 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd2
Genomic Map